Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29323952

Superose® 6 Prep Grade HiLoad®

column thread (16/600), Cytiva 29323952

Synonym(s):

Superose 6 Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NB.21

feature

Sample volume less than 5 mL

manufacturer/tradename

Cytiva 29323952

parameter

flow rate (1.6 mL/min)

bed volume × D × L

(120 mL) (16 mm) (600 mm)

column I.D.

(16 mm)

column thread

(16/600)

General description

HiLoad 16/600 Superose 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 mL.


Selectivity for a wide range of proteins: This column is packed with Superose 6 pg resin, which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5 000 000.

Features and Benefits

  • Convenience and reproducibility: can be used with standalone pump or a chromatography system.
  • High-resolution separations: purification is possible across an exceptionally broad molecular-weight range.
  • Scalable – separations can be scaled up using Superose 6 prep grade resin.
  • ÄKTA system compatible – Fittings (1/16”) for direct connection to ÄKTA systems and other chromatography systems.

Legal Information

HiLoad is a registered trademark of Cytiva
Superose is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service