Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE28-9323-61

Vivaspin® 20, 30 kDa MWCO Polyethersulfone

Cytiva 28-9323-61, pack of 12 pieces

Synonym(s):

30 kDa Vivaspin, Vivaspin 20 MWCO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

material

polycarbonate housing
polyethersulfone membrane

sterility

non-sterile

packaging

pack of 12 pieces

manufacturer/tradename

Cytiva 28-9323-61

availability

not available in Japan

tube size

50 mL , conical bottom

working volume

20 mL

pore size

30 kDa MWCO

storage temp.

room temp

General description

Designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%.

Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cut-off (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.

Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3 000 to 100 000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest

Features and Benefits

  • Compatible pH range is from pH 1 to 9.
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss.
  • Vertical polyethersulfone membrane minimizes membrane blockage and tolerates high flow rates.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Vivaspin is a registered trademark of Sartorius Stedim Biotech GmbH

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Diep et al.
Analytical biochemistry, 609, 113836-113836 (2020-08-05)
Solute-binding proteins (SBPs) from ATP-binding cassette (ABC) transporters play crucial roles across all forms of life in transporting compounds against chemical gradients. Some SBPs have evolved to scavenge metal substrates from the environment with nanomolar and micromolar affinities (KD). There

Articles

This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.

Protocols

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service