Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9180-08

PD Miditrap G-25

Cytiva 28-9180-08

Synonym(s):

G-25 Miditrap, Miditrap G-25, PD G-25 Miditrap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NA.56

material

polypropylene and polyethylene resin
resin (Sephadex G-25)

sterility

non-sterile

feature

wetted part: no

packaging

pkg of 50 ea (columns)

manufacturer/tradename

Cytiva 28-9180-08

storage condition

dry media

bed size

13 mm × 26 mm

bed volume

3.5 mL

column I.D.

13 mm

working volume

0.5—1 mL

matrix

cross-linked dextran

particle size

>50 μm (dry)
85-260 μm

pore size

5000  Da MWCO

cleaning

2-13

working range

2-13

suitability

suitable for bioprocess medium

Related Categories

General description

PD MidiTrap G-25 is a convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel. Can be used in a centrifugation or gravity protocol.

PD MidiTrap G-25 is designed for convenient desalting and buffer exchange of biological samples and volumes up to 1.0 mL can be handled. Two alternative application protocols are available, gravity or centrifugation. By using the gravity protocol, a simple cleanup of the sample is done with one or several columns in parallel without any need for a purification system. With the centrifugation protocol, the samples are run in parallel in a standard centrifuge with minimal dilution of the eluted sample. Four adapters are included in each product pack to enable easy centrifugation.

Application

Protein sample preparation. Desalting/Buffer Exchange/Cleanup.

Features and Benefits

  • Reliable and reproducible cleanup of proteins and other large biomolecules (>5000 Mr).
  • Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds.
  • High desalting capacity.
  • Flexible protocols with ability to use gravity flow or centrifugation.
  • Sample preparation in the range of 0.5 to 1.0 mL for gravity and 0.75 to 1.0 mL with centrifugation.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.15% Kathon CG/ICP Biocide)
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (high sub) media and columns from Cytiva.

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

Protocols

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service