Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9042-58

illustra Bacteria Mini Spin

Cytiva 28-9042-58, sufficient for 50 preparations

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 50 preparations

packaging

pkg of 50 ea

manufacturer/tradename

Cytiva 28-9042-58

General description

Bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.

Most bacteria have a genome that consists of a single DNA molecule (i.e., one chromosome that is several million base pairs in size and is ""circular"": without ends like the chromosomes of eukaryotic organisms). In addition, bacteria may have one or more smaller circular DNA molecules, called plasmids, that usually contain non-essential genes.

Genomic DNA is found in the chromosomes and it is much longer and higher in molecular weight than plasmid DNA. Genomic DNA is the code for the genome, and this is what is needed to do genome sequencing, and for applications such as bacterial genotyping and microbiome sequencing.

For sequencing it is desirable that the bacterial DNA extraction methods used allow the collection of intact genomic DNA in the longest fragments possible. A genomic DNA kit format can help standardize the procedures used to prepare bacterial genomic DNA samples for further analysis.

Bacteria genomicPrep Mini Spin Kit can be used for genomic DNA extraction from a wide range of bacterial strains such as DH5α, TOP10, JM109, Bacillus subtilis, etc.

The kit produces highly pure and intact genomic DNA with A260/A280 values of 1.75 to 1.85. This provides DNA at concentrations suitable for downstream applications such as bacterial genotyping and microbiome testing for bacteria identification, as well as more standard techniques such as restriction enzyme digestion, RT-PCR, and long PCR.

The method uses a lysis solution in combination with proteinase K to gently release genomic DNA into solution from bacterial cells. The DNA is deproteinated in the extraction solution and the genomic DNA is then bound onto a silica column in the presence of chaotropic solution. Contaminants are removed during wash steps and the purified DNA is eluted with prewarmed TE solution.

Features and Benefits

  • For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteria.
  • Total DNA extraction time under 1 h with reduced lysis time.
  • Can be used to purify 4 to 8 µg of genomic DNA from 2 × 1NA9 bacteria cells with input amounts ranging from 1 × 1NA9 to 4 × 1NA9 bacteria cells.
  • Spin columns, prepacked with a silica membrane, provide excellent reproducibility and consistency.
  • Verified in several downstream applications including RT PCR, long PCR, and restriction enzyme digests.
  • Color-coded caps for ease of use.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows downstream applications after genomic DNA purification.

Description of common problems that can occur during genomic DNA preparation

Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.

Protocols

This page shows Genomic DNA purification using illustra™ genomicPrep Mini Spin Kits from Cytiva.

This page shows genomic DNA purification from sample on FTA elute from Cytiva.

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service