Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-449

Sigma-Aldrich

Anti-Bin1 Antibody, clone 99D

clone 99D, Upstate®, from mouse

Synonym(s):

Anti-AMPH2, Anti-AMPHL, Anti-CNM2, Anti-SH3P9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

99D, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BIN1(274)
mouse ... Bin1(30948)

Specificity

Bin1

Immunogen

Partial fusion protein corresponding to amino acids 189-398 of human Bin1. Clone 99D

Application

Anti-Bin1 Antibody, clone 99D is an antibody against Bin1 for use in IP, WB, IH.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Quality

routinely evaluated by immunoblot on RIPA lysate of C2C12 cells.

Target description

56kDa

Linkage

Replaces: MAB4131

Physical form

0.1M Tris-glycine, pH 7.4, containing 0.05% sodium azide
Format: Purified
Protein G Chromatography

Storage and Stability

1 year at -20°C

Other Notes

Due to license agreement restrictions, this product cannot be purchased for resale.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Wechsler-Reya et al.
Cancer research, 57(15), 3258-3263 (1997-08-01)
BIN1 is a putative tumor suppressor that was identified in a genetic screen for polypeptides that interact with the MYC oncoprotein. Using a set of six monoclonal antibodies, we identified and examined biochemical features and localization of cellular BIN1. Epitope
Sisi Yang et al.
Neuron, 110(5), 824-840 (2022-01-24)
Autophagy is a cellular degradation pathway essential for neuronal health and function. Autophagosome biogenesis occurs at synapses, is locally regulated, and increases in response to neuronal activity. The mechanisms that couple autophagosome biogenesis to synaptic activity remain unknown. In this
Stephanie L Adams et al.
Journal of neuropathology and experimental neurology, 75(8), 779-790 (2016-10-22)
Genome-wide association studies have established BIN1 (Bridging Integrator 1) as the most significant late-onset Alzheimer disease (AD) susceptibility locus after APOE We analyzed BIN1 protein expression using automated immunohistochemistry on the hippocampal CA1 region in 19 patients with either no
Suppression of MEK/ERK signalling by Myc: role of Bin-1.
Telfer, JF; Urquhart, J; Crouch, DH
Cellular Signalling null
R J Wechsler-Reya et al.
Molecular and cellular biology, 18(1), 566-575 (1998-01-07)
Bin1 is a Myc-interacting protein with features of a tumor suppressor. The high level of Bin1 expression in skeletal muscle prompted us to investigate its role in muscle differentiation. Significant levels of Bin1 were observed in undifferentiated C2C12 myoblasts, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service