Key Documents
GE18-1004-30
Immobiline® DryStrip kit, for Immobiline DryStrip gels with MultiPhor II Electrophoresis System
pack of 1 kit
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
feature
wetted part: no
packaging
pack of 1 kit
manufacturer/tradename
Cytiva 18100430
General description
This kit is designed for running the first dimension of 2-D electrophoresis using precast Immobiline DryStrip gels, see Immobiline DryStrip Gels. You can apply up to 1NANA µl of sample per strip and run up to 12 strips simultaneously
This Instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment. Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps. IPG also makes it possible to focus basic proteins in the gel and to obtain distinct protein spots. Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
Features and Benefits
  • For use of immobiline DryStrip gels with Multiphor II electrophoresis system.
Legal Information
Immobiline is a registered trademark of Cytiva
MultiPhor is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This protocol shows how to prepare the Immobiline® DryStrip Kit and electrodes strips.