Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE17-5279-02

Glutathione Seph High Performance

Cytiva 17-5279-02, pack of 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

glutathione

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-5279-02

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

3-12

working range

3-12

capacity

>7 mg/mL binding capacity(medium)

General description

Glutathione Sepharose High Performance is an affinity medium for high resolution purification of Glutathione S transferase (GST) tagged proteins.

Application

Glutathione Sepharose High Performance is used for the purification of GST-tagged Proteins from bacterial lysates and other S-transferases or glutathione-dependent Proteins. GST-tagged Proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves Protein antigenicity and function.

Features and Benefits

  • 34 μm bead size for improved efficiency and resolution
  • Fast, one-step purification of GST-tagged Proteins and other glutathione S-trasferase or glutathione-dependent Proteins
  • Mild conditions during the purification process preserves Protein antigenicity and function

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes general considerations for purification of GST-tagged proteins using Cytiva products.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

See All

Protocols

This page shows how to purify GST-tagged proteins using Glutathione Sepharose from Cytiva.

GSTrap™ affinity columns are specially designed 1 ml and 5 ml HiTrap® columns packed with Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, or Glutathione Sepharose 4B media.

GSTrap™ affinity columns facilitate protein purification with Glutathione Sepharose® chromatography media for efficient purification processes.

GSTrap™ affinity columns facilitate protein purification with Glutathione Sepharose® chromatography media for efficient purification processes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service