Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0956-01

Gelatin-Sepharose 4B

Cytiva 17-0956-01, pack of 25 mL

Synonym(s):

Capto DEAE HiTrap, DEAE HiTrap Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

bovine gelatin

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-0956-01

storage condition

(20% Ehtanol)

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

3-12

working range

3-10

storage temp.

2-8°C

General description

Gelatin Sepharose 4B affinity media with immobilized gelatin is designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificity. Gelatin Sepharose 4B is bovine gelatin coupled to Sepharose 4B by the cyanogen bromide method. Gelatin binds specifically to fibronectin, which is a high molecular weight glycoprotein found on the surfaces of many cell types and present in many extracellular fluids including plasma. Gelatin Sepharose™ 4B has been designed for the purification or removal of fibronectin.

Application

Gelatin binds specifically to fibronectin, which is a high molecular weight glycoProtein found on the surfaces of many cell types and present in many extracellular fluids including plasma. Gelatin Sepharose 4B has been designed for the purification or removal of fibronectin.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service