Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0886-01

Gammabind Plus Sepharose

Cytiva 17-0886-01, pack of 5 mL

Synonym(s):

Affinity chromatography resin, Protein A Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

recombinant GammaBind G type 3 lacking albumin-binding region

packaging

pack of 5 mL

manufacturer/tradename

Cytiva 17-0886-01

storage condition

(20% Ehtanol, PBS pH 7)

matrix

6% cross-linked agarose

average diameter

90 μm

cleaning in place

2-9

working range

3-9

shipped in

wet ice

storage temp.

2-8°C

General description

GammaBind Plus Sepharose is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to GammaBind Sepharose.

GammaBind Plus Sepharose is GammaBind G, Type 3, covalently immobilized to Sepharose CL-6B by malimide linkage. GammaBind G, Type 3, a recombinant form of streptococcal protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale.
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse.
  • Yields up to 18 mg/mL mouse IgG.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service