Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8644

Sigma-Aldrich

D-(+)-Glucose solution

100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

D-Glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1281608
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

product line

BioXtra

form

solution

concentration

100 g/L in H2O

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

Glucose provides the primary energy source for cell metabolism. It is a main source of energy for living organisms. Glucose occurs naturally in the free state in fruits and other parts of plants. It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.

Application

D-(+)-Glucose solution has been used:

  • to determine extracellular acidification rate (ECAR) or oxygen consumption rate (OCR) in human CB CD34+cells by Seahorse extracellular flux assay
  • as a negative control in human umbilical vein endothelial cell (HUVEC) culture
  • as a supplement for culturing colonic biopsies

Biochem/physiol Actions

Glucose is an aldosic monosaccharide and acts as an important component of glycolysis and downstream pathways for aerobic and anaerobic respiration. It provides energy for growth and reproduction. Glucose is a byproduct of photosynthesis that is condensed to starch in plants and cyanobacteria.

Other Notes

This product has been tested with cell lines to verify the product is not cytotoxic. The growth promoting capacity of this cell culture tested glucose solution was assessed at 45 mL/L in medium using appropriate cell lines.
This solution has been sterilized by filtration.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu J Lam et al.
ACS applied materials & interfaces, 8(49), 33446-33456 (2016-12-15)
'Structurally nanoengineered antimicrobial peptide polymers' (SNAPPs), in the form of star-shaped peptide polymer nanoparticles, have been recently demonstrated as a new class of antimicrobial agents with superior in vitro and in vivo efficacy against Gram-negative pathogens, including multidrug-resistant species. Herein
Brandon Mapes et al.
Physiological genomics, 46(8), 302-308 (2014-02-20)
1α,25-Dihydroxyvitamin D3 [1α,25(OH)2D3] is a steroid hormone derived from circulating 25(OH) vitamin D [25(OH)D] with chemopreventive effects in colorectal cancer. 1α,25(OH)2D3 acts through transcriptional mechanisms; however, our understanding of vitamin D transcriptional responses in the colon is derived from studies
Qianqian Li et al.
Journal of cellular and molecular medicine, 24(1), 910-920 (2019-11-05)
In type 1 and type 2 diabetes mellitus, increased cardiac fibrosis, stiffness and associated diastolic dysfunction may be the earliest pathological phenomena in diabetic cardiomyopathy. Endothelial-mesenchymal transition (EndMT) in endothelia cells (ECs) is a critical cellular phenomenon that increases cardiac
Dieter Henrik Heiland et al.
Nature communications, 10(1), 2541-2541 (2019-06-13)
Reactive astrocytes evolve after brain injury, inflammatory and degenerative diseases, whereby they undergo transcriptomic re-programming. In malignant brain tumors, their function and crosstalk to other components of the environment is poorly understood. Here we report a distinct transcriptional phenotype of
Andreia V Pinho et al.
PloS one, 10(6), e0128012-e0128012 (2015-06-06)
Sirtuin 1 (Sirt1) has been reported to be a critical positive regulator of glucose-stimulated insulin secretion in pancreatic beta-cells. The effects on islet cells and blood glucose levels when Sirt1 is deleted specifically in the pancreas are still unclear. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service