Skip to Content
MilliporeSigma

Skip To

EC810

GenElute -E Viral RNA/DNA Swab Kit

Reagents and consumables for viral RNA and DNA purification. 10, 50 or 250 purifications.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 reactions
Check Cart for Availability
$301.00
250 reactions
Check Cart for Availability
$1,340.00

About This Item

UNSPSC Code:
41105501
NACRES:
NB.23

$301.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

technique(s)

DNA purification: suitable, RNA purification: suitable

input

sample(s) (swabs: nasopharyngeal and genital)
sample(s) (stool)

test parameters

: 3 min hands on time, sample volume: 10-20 mg stool, sample volume: 50 μL swabs

storage temp.

2-8°C

General description

GenElute-E purification kits provide a simple alternative for viral RNA and DNA purification. Compared with traditionalmethods, GenElute-E offers:
  • Isolation of viral RNA and DNA in less than 30 minutes with minimal sample handling
  • Better purity, leading to improved performance in PCR and other applications
  • Significant reduction in plastic and hazardous chemical waste

Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, and subject the nucleic acids to centrifugation sheering forces.

GenElute-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in swab, cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminantswhile allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-qualitypurification is made possible by our innovative SmartLyse® Buffer, which enables fast and efficient isolation.

Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply prepare spin column, mix sample with lysis components, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified viral RNA and DNA is ready to be used in downstream applications or stored.

Application

Suitable for most common downstream applications, including genotyping, PCR, and NGS
The GenElute-E Viral RNA/DNA Swab kit has been developed to purify genomic viral RNA and DNA from nasopharyngeal swabs, dry or in transport medium, in addition to genital swabs and stool samples.

The sample lysis utilizes the SmartLyse® Viral Buffer for fast and efficient isolation of nucleic acids from a variety of swab types and stool samples.

Purified viral RNA and DNA is eluted in Tris buffer, pH 7.8 and can immediately be used for most common downstream applications, including PCR, genotyping, NGS, and others. Final yield is comparable to most common silica-based methods. GenElute-E purified genomic DNA preparations commonly show an A260/280 ratio of around 1.8.

Features and Benefits

  • Fast and efficient purification: no lysis incubation and minimal hands-on, preparation steps
  • Higher quality nucleic acids: negative chromotography-based purification reduces carryover of PCR inhibitors and other contaminents
  • Reduced environmental impact: 55% less plastic waste, as well as a significant reduction in hazardous chemicals

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised.  The single-spin protocol limits the number of collection and pipetting steps, reducing plastic waste by 55% compared to equivalent bind-wash-elute methods. GenElute-E is designated a Greener Alternative Product based on the ""Prevention"" and ""Designing Safer Chemicals"" qualifications of The 12 Principles of Green Chemistry.GenElute-E kits also adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
SmartLyse is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EC848EC896EC996
technique(s)

DNA purification: suitable, RNA purification: suitable

technique(s)

-

technique(s)

-

technique(s)

-

test parameters

: 3 min hands on time, sample volume: 50 μL swabs, sample volume: 10-20 mg stool

test parameters

-

test parameters

-

test parameters

-

input

sample(s) (swabs: nasopharyngeal and genital)
sample(s) (stool)

input

DNA
RNA

input

DNA
RNA

input

RNA
DNA

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp


Kit Components Only

Product No.
Description

  • SmartLyse® Viral Buffer

  • GenElute-E Spin Columns



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. Genelute-e single spin DNA and RNA purification kits use negative chromatographty to isolate nucleic acids. Can you explain how this approach simplifies workflows?

    1 answer
    1. Instead of optimizing the bind, wash, and release steps in conventional silica-based spin purification preps, the technology focuses on a separation by performing a single step fractionation based on the size of the biomolecules, which results in depleted impurities.

      Helpful?

  2. Do we know how stable the purified DNA is through several freeze-thaw cycles?

    1 answer
    1. This will fluctuate due to sample variability (sample collection, concentration, fragment length, sequence [GC content], storage before isolation, etc.). However, the sample is buffer exchanged into a standard storage buffer that is included in the kit (1X TE).

      Helpful?

  3. What is the composition of the lysis buffer and clearing buffer after flowing through the resin?

    1 answer
    1. The presence of EDTA, SDS, or excess salt can affect my PCR/ sequencing reaction. The lysis buffer information is proprietary, but we can say it is free of chaotropic salts. The resins are desalting resins so EDTA, SDS, and salts are depleted.

      Helpful?

  4. Does the technology introduce any bias into the sample?

    1 answer
    1. GenElute™-E does not introduce biases that some “bind-wash-elute” technologies can add because the technology separates by size, rather than by what binds and what is released.

      Helpful?

Reviews

No rating value

Active Filters