Skip to Content
MilliporeSigma
All Photos(3)

Documents

G7375

Sigma-Aldrich

D-Glucose 6-phosphate dipotassium salt hydrate

Sigma Grade, 98-100% dry basis (enzymatic)

Synonym(s):

D(+)-Glucopyranose 6-phosphate dipotassium salt, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11K2O9P · xH2O
CAS Number:
Molecular Weight:
336.32 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

grade

Sigma Grade

Assay

98-100% dry basis (enzymatic)

form

powder

impurities

<4 methanol (GC)

color

white

cation traces

K: 20.4 -26.1% (dry basis)
P: <0.1%

storage temp.

−20°C

SMILES string

[K].OC(COP(O)(O)=O)C(O)C(O)C(O)C=O

InChI

1S/C6H13O9P.K.H/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;;/h1,3-6,8-11H,2H2,(H2,12,13,14);;

InChI key

OSJCIPGPFUZJML-UHFFFAOYSA-N

General description

Glucose is converted to D-Glucose 6-phosphate by phosphorylation of the 6′ carbon by either hexokinase or glucokinase in liver cells. D-Glucose 6-phosphate can be stored in the liver or muscle tiβue as glycogen until the body needs additional glucose where it will be proceβed by glycogen phosphorylase and released.

Quality

usually 2-4 H2O/mole

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berg, J. M.
Biochemistry (5th Edition) (2002)

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service