Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G211600

Sigma-Aldrich

DMT-2′O-TC-rG(ib) Phosphoramidite

Synonym(s):

N2-Isobutyryl-O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)guanosine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H61N8O11PS2
CAS Number:
Molecular Weight:
1033.16
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

solid

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)
≤0.5% 3′-TC isomer (reversed phase HPLC)
≤0.5% P(III) 145-155ppm (31P-NMR)

color

white to light yellow

solubility

acetonitrile: toluene (1:1): 0.1 M at 18 °C, clear (for 72h)

λ

conforms (UV/VIS Identity)

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: TC
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](OC(=S)N3CCS(=O)(=O)CC3)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n4cnc5C(=O)NC(NC(=O)C(C)C)=Nc45)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C49H61N8O11PS2/c1-31(2)44(58)53-47-52-43-40(45(59)54-47)51-30-56(43)46-42(67-48(70)55-24-27-71(60,61)28-25-55)41(68-69(65-26-12-23-50)57(32(3)4)33(5)6)39(66-46)29-64-49(34-13-10-9-11-14-34,35-15-19-37(62-7)20-16-35)36-17-21-38(63-8)22-18-36/h9-11,13-22,30-33,39,41-42,46H,12,24-29H2,1-8H3,(H2,52,53,54,58,59)/t39-,41-,42-,46-,69?/m1/s1

InChI key

LXUNLMGCWYNULU-NWGULGMGSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

For use with Customer Container sold to end user customers either directly or through distributors.

This product is manufactured and sold under a license from Agilent Technologies under under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies. The purchase of this product conveys to the buyer the limited, non-exclusive, non-transferable right (without the right to resell, repackage, or further sublicense) under these patents to use the product to synthesize Oligonucleotides for buyer′s internal research use only, including commercial purposes. Except as otherwise provided herein, no other license is granted to the buyer weather expressly, by implication, by estoppel or otherwise. Buyer agrees and understands that it is not licensed to use this product to synthesize Oligonucleotides for sale, for diagnostic uses including use in clinical diagnostic settings where the use in conjunction with an individual′s sample generates data provided to an individual or the individual′s caregiver for a free reimbursement, neither can Oligonucleotides synthesized using this product be used for use clinical trials or for other therapeutic purposes. This product is sold pursuant to authorization from Agilent Technologies, Inc. and Agilent Technologies reserves all other rights under these patents.

For use with all sales where the purchaser is required to have its own direct license from Agilent, without limitation, Bulk Quantity Sales, and sales to Oligo Houses.

This product and it uses are covered by under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies. This product is sold to purchaser pursuant to authorization by Agilent Technologies, Inc. without any rights conferred by seller, based on purchaser′s direct license and authorization to purchase from Agilent Technologies, Inc. No license is granted to the purchaser weather expressly, by implication, by estoppel or otherwise on account of this purchase. Purchase′s rights to use this product are governed solely and expressly by purchaser′s direct license agreement with Agilent.
These products are manufactured and sold under a license from Agilent Technologies under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies.

These products and its uses are covered by US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service