Skip to Content
MilliporeSigma

G1875

β-Galactosidase from bovine liver

Grade III, lyophilized powder, ≥0.15 units/mg protein

Synonym(s):

β-D-Galactoside galactohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-864-1
MDL number:
EC Number:
Specific activity:
≥0.15 units/mg protein
Biological source:
bovine liver
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine liver

Quality Segment

type

Grade III

form

lyophilized powder

specific activity

≥0.15 units/mg protein

composition

Protein, ≥40%

shipped in

wet ice

storage temp.

−20°C

Application

β-galactosidase was used in the production of a stabilized, single reagent for alcohol analysis.

Biochem/physiol Actions

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Other Notes

One unit will hydrolyze 1.0 μmole of o-nitrophenyl β-D-galactoside to o-nitrophenol and D-galactose per min at pH 7.3 at 37 °C.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library