Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP951040227

Eppendorf® Microplate 96/V-PP

sterile, white border, clear Wells, with conical bottom, pkg of 80 plates (5 bags x 16 plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
material:
colorless wells
conical bottom wells
size:
96 well
sterility:
sterile

material

colorless wells
conical bottom wells

sterility

sterile

packaging

pkg of 80 plates (5 bags x 16 plates)

manufacturer/tradename

Eppendorf® 951040227 (North America)

size

96 well

color

clear Wells, with conical bottom
white border

suitability

suitable for PCR
suitable for manual and automated sequencing techniques

Related Categories

General description

The “Eppendorf” Microplate 96/V-PP is a specially designed polypropylene microplate consisting of 96 wells. These microplates have significantly improved the process of pipetting, controlling, and recovering of samples.

Application

The “Eppendorf” Microplate 96/V-PP may be used for:
  • Sample preparation and storage
  • Biological assays which require a high resistance against temperature or solvents
  • Screening of active ingredients
  • Storage of nucleotide libraries

Features and Benefits

  • The microplates are made from high quality polypropylene, ensuring an improved sample visibility.
  • Polypropylene is highly resistant to chemicals, mechanical stressors, and extreme temperature conditions.
  • The [TM=“Eppendorf”] Microplate 96/V-PP demonstrates exceptional centrifugation stability up to 6,000 × g.
  • The [TM=“OptiTrack”] Matrix feature has fastened up sample identification and reduced pipetting errors.
  • An optimized [TM=“RecoverMax”] well geometry minimizes the remaining volume and promotes excellent mixing properties.
  • Elevated well rims and a smooth surface, ensures effective sealing.
  • The [TM=“Eppendorf”] Microplate 96/V-PP functions across a wide temperature range of -86 °C to 100 °C.
  • They can be Autoclaved at 121 °C for 20 min.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service