Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96M2HSC

PCR amplification plate microplate

half skirt with a single notch, size 96 wells, clear, pkg of 5x10plates/cs

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
well volume:
200 μL

material

conical bottom wells

sterility

non-sterile

feature

lid: no
skirt
Half skirt

manufacturer/tradename

Axygen Inc PCR-96M2-HS-C
Corning

packaging

pkg of 5x10plates/cs

size

96 wells

well volume

200 μL

color

clear

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® 96 well half skirt PCR microplate is ideal for use with standard thermal cycling, plus ABI PCR 2700, 2720, 9700, 9800, 7300, 7500, 7900 and ABI sequencing units systems.
  • Uniform ultra-thin wall thickness facilitates optimal thermal transfer.
  • Free of DNase-/RNase and nonpyrogenic.
  • Sealing options : Strip caps, Axymat, or sealing film.
  • Single notch corner.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service