Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5910758006

Eppendorf® Adapter

12 x 75 mm for S-4xUniversal, pack of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

12 x 75 mm for S-4xUniversal
Holds 5 mL FACS tubes

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5910758006

technique(s)

centrifugation: suitable

compatibility

for use with Universal bucket

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service