Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5920746006

Eppendorf® Adapter for round-bottom tubes

17.5 x 100 mm., for rotor S-4xUniversal-Large

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82

feature

17.5 x 100 mm., for rotor S-4xUniversal-Large
Holds 9 mL tubes

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5920746006

technique(s)

centrifugation: suitable

compatibility

for use with Universal-Large bucket

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service