Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP4423000010

Eppendorf® Pipet Helper®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

mechanical
pipetting type: air-cushion

manufacturer/tradename

Eppendorf® 4423000010 (Global)

volume

100 mL

General description

The Eppendorf Pipet Helper in additon, is the perfect instrument for inexperienced users as its robust and intuitive design allows everybody to adjust the meniscus precisely. In addition the valve unit is optimized so that liquids can be drawn up simply, without exerting pressure. The aspiration bulb provides rapid filling of the pipet. The aspiration and dispensing of liquids are controlled gently by the ergonomic lever. Additionally a splash free blow-out is provided. The specially designed aspirating cone ensures secure seating for all measuring and volumetric pipets (glass and plastic).

Application

Preparation of a buffer for chromatography
Preparation of a fluid for fermentation
Sample transfer within petrochemistry
Generate a sample within water analytiks
Preparation of assays

Features and Benefits

Lightweight with reduced operating force
Easy maintenance and cleaning due to easy disassembly, few parts and fully autoclavable
Comfortable working because of the good balance and pleasing material
The aspirating bulb enables pipetting of volumes up to 100 mL at once
Single lever controls low force aspirating and dispensing
High control over the liquid movement due to very sensitive valve unit
Drop wise dispensing and blow out without splashing
Drainage without any additional pressure delivers very reproducible results
Drip stop via hydrophobic membrane filter (3 μm)
The special silicon pipet adapter guarantees a secure pipette fit

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service