Skip to Content
MilliporeSigma
All Photos(1)

Documents

EP022639905

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

material

aluminum

feature

RCF 3,214 × g

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022639905

compatibility

5804/5804R & 5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

Rotor S-4-104, including software upgrade (required for some older models)
  • Medium capacity swing-bucket rotor with max. speed 3,214 × g (3,900 rpm)1)
  • Broad range of adapters enables high versatility for tubes and bottles from 0.2 mL to 750 mL and plates
  • Aerodynamic round bucket design for up to 36 % energy savings compared to Rotor A-4-81
  • Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility
  • Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available2)
  • Buckets, lids and adapters are autoclavable (121 °C, 20 min)

1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2)Aerosol-tightness tested by Public Health England, Porton Down, UK.

Features and Benefits

Medium capacity swing-bucket rotor with max. speed 3,214 × g (3,900 rpm)1)Broad range of adapters enables high versatility for tubes and bottles from 0.2 mL to 750 mL and platesAerodynamic round bucket design for up to 36 % energy savings compared to Rotor A-4-81Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatilityAerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available2)Buckets, lids and adapters are autoclavable (121 °C, 20 min)1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2)Aerosol-tightness tested by Public Health England, Porton Down, UK.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service