Key Documents
EP0030108418
Eppendorf® DNA LoBind Tubes
capacity 1.5 mL, PCR clean, pack of 500 ea
Synonym(s):
Eppendorf® DNA LoBind tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material
cap (push fit)
colorless
conical bottom
description
Low retention
withstand, 30,000 x g
sterility
non-sterile
feature
DNase free
PCR clean
packaging
pack of 500 ea
capacity
1.5 mL
suitability
suitable for PCR
Features and Benefits
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules
Legal Information
Eppendorf is a registered trademark of Eppendorf AG