EP0030108400
Eppendorf® DNA LoBind Tubes
capacity 0.5 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Synonym(s):
Eppendorf® DNA LoBind tubes
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
cap (push fit)
colorless
conical bottom
description
Low retention
sterility
non-sterile
feature
DNase free
PCR clean
packaging
pack of 500 ea
capacity
0.5 mL
suitability
suitable for PCR
Related Categories
General description
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids
Features and Benefits
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)