Skip to Content
MilliporeSigma

E2535

Eprosartan mesylate

≥98% (HPLC)

Synonym(s):

Eprosartan Monomethanesulfonate, SKF-108566J, Teveten

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H24N2O4S · CH4O3S
CAS Number:
Molecular Weight:
520.62
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Abbott

storage temp.

room temp

SMILES string

CS(O)(=O)=O.CCCCc1ncc(\C=C(/Cc2cccs2)C(O)=O)n1Cc3ccc(cc3)C(O)=O

InChI

1S/C23H24N2O4S.CH4O3S/c1-2-3-6-21-24-14-19(12-18(23(28)29)13-20-5-4-11-30-20)25(21)15-16-7-9-17(10-8-16)22(26)27;1-5(2,3)4/h4-5,7-12,14H,2-3,6,13,15H2,1H3,(H,26,27)(H,28,29);1H3,(H,2,3,4)/b18-12+;

InChI key

DJSLTDBPKHORNY-XMMWENQYSA-N

Gene Information

human ... AGTR1(185)

Biochem/physiol Actions

Angiotensin II type 1 (AT1) receptor antagonist; anti-hypertensive.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Eprosartan mesylate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library