
Eprosartan mesylate

Synonym(s):
Eprosartan Monomethanesulfonate, SKF-108566J, Teveten
Empirical Formula (Hill Notation):
C23H24N2O4S · CH4O3S
CAS Number:
Molecular Weight:
520.62
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing