Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D8555

Sigma-Aldrich

N,N-Dihexyl-2-(4-fluorophenyl)indole-3-acetamide

Synonym(s):

FGIN-1-27

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37FN2O
CAS Number:
Molecular Weight:
436.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

solubility

methanol: 50 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCCCCCN(CCCCCC)C(=O)Cc1c([nH]c2ccccc12)-c3ccc(F)cc3

InChI

1S/C28H37FN2O/c1-3-5-7-11-19-31(20-12-8-6-4-2)27(32)21-25-24-13-9-10-14-26(24)30-28(25)22-15-17-23(29)18-16-22/h9-10,13-18,30H,3-8,11-12,19-21H2,1-2H3

InChI key

VUWXAQFLTSBUDB-UHFFFAOYSA-N

Gene Information

human ... BZRAP1(9256)
rat ... Tspo(24230)

Biochem/physiol Actions

High affinity ligand for glial mitochondrial benzodiazepine receptors; stimulates neurosteroid biosynthesis; does not bind GABAA receptors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dandan Zhou et al.
Journal of molecular neuroscience : MN, 70(4), 542-549 (2019-12-28)
Microglia activation has been reported to be associated with pathogenesis of neuroinflammation, central nervous system damage, and degeneration diseases. With various damage-associated molecules released, M1 polarization of microglia emerges early after injury and followed by M2 polarization. In this study
E Romeo et al.
The Journal of pharmacology and experimental therapeutics, 267(1), 462-471 (1993-10-01)
The 2-aryl-indole-3-acetamide derivatives, 2-hexyl-indole-3-acetamide (FGIN-1-27) and 2-hexyl-indole-3-acetamide-N-benzene-tricarboxylic acid (FGIN-1-44) displaced [3H]1-(2-chlorophenyl)-N-methyl-N-(1- methylpropyl)-3-isoquinoline-carboxamide([3H]PK 11195) and [3H]4-chlorodiazepam ([3H]4'CD) from binding sites located on the rat brain mitochondrial DBI receptor complex (MDRC) with Ki values in the nanomolar range. Both 2-aryl-indole-3-acetamide derivatives acted
F M Freeman et al.
Pharmacology, biochemistry, and behavior, 67(2), 355-362 (2000-12-22)
The specific mitochondrial benzodiazepine receptor (MBR) agonist, FGIN 1-27, and antagonist, PK 11195, were used to investigate whether this receptor was involved in passive avoidance memory formation in the day-old chick. PK 11195 at a concentration of 1-10 microM was
Kazuhiro Tokuda et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(50), 16788-16795 (2010-12-17)
Benzodiazepines (BDZs) enhance GABA(A) receptor inhibition by direct actions on central BDZ receptors (CBRs). Although some BDZs also bind mitochondrial receptors [translocator protein (18 kDa) (TSPO)] and promote the synthesis of GABA-enhancing neurosteroids, the role of neurosteroids in the clinical
Cheryl A Frye et al.
Neuroendocrinology, 77(1), 71-82 (2003-03-08)
Progestins modulate lordosis through actions in the ventral tegmental area (VTA). Whether neurosteroidogenesis of 5alpha-pregnan-3alpha-ol-20-one (3alpha,5alpha-THP), involving mitochondrial benzodiazepine receptors (MBR), is important for lordosis was investigated. Ovariectomized (Ovx), hormone-primed rats (experiments 1, 3, 5, 6) and rats in behavioral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service