Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D5297

Sigma-Aldrich

Dehydroisoandrosterone 3-sulfate sodium salt dihydrate

≥93% (TLC), powder

Synonym(s):

5-Androsten-3β-ol-17-one sulfate sodium salt dihydrate, Prasterone sulfate sodium salt dihydrate, Sodium dehydroepiandrosterone sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NaO5S · 2H2O
CAS Number:
Molecular Weight:
426.50
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥93% (TLC)

form

powder

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to off-white

mp

148-149 °C (dec.) (lit.)

solubility

methanol: soluble, clear to hazy

SMILES string

O.O.[Na+].C[C@]12CC[C@H]3[C@@H](CC=C4C[C@H](CC[C@]34C)OS([O-])(=O)=O)[C@@H]1CCC2=O

InChI

1S/C19H28O5S.Na.2H2O/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;;;/h3,13-16H,4-11H2,1-2H3,(H,21,22,23);;2*1H2/q;+1;;/p-1/t13-,14-,15-,16-,18-,19-;;;/m0.../s1

InChI key

NLNMKDUYGPNWAO-OXNWJOIVSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Dehydroisoandrosterone 3-sulfate sodium salt dihydrate has been used as a supplement to induce superoxide anion production in human neutrophil granulocytes.

Biochem/physiol Actions

Dehydroisoandrosterone 3-sulfate sodium salt dihydrate is a negative allosteric modulator of GABAA receptors.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma concentration of myeloperoxidase enzyme in pre-and post-climacterial people: related superoxide anion generation
Bekesi G, et al.
Experimental Gerontology, 37(1), 137-148 (2001)
S L Hansen et al.
Molecular pharmacology, 55(3), 489-496 (1999-03-03)
Dehydroepiandrosterone sulfate (DHEAS) is a neuroactive steroid with antagonist action at gamma-aminobutyric acid type A (GABAA) receptors. Patch-clamp techniques were used to investigate DHEAS actions at GABAA receptors of the rat pituitary gland at two distinct loci: posterior pituitary nerve
Manuel de Jesús Gallegos-Saucedo et al.
Neurotoxicity research, 38(2), 447-460 (2020-05-16)
Several pathophysiological processes involve Hypoxia conditions, where the nervous system is affected as well. We postulate that the GABAergic system is especially sensitive. Furthermore, drugs improving the resistance to hypoxia have been investigated, such as the neurosteroid dehydroepiandrosterone sulfate (DHEAS)
P A Lapchak et al.
Stroke, 31(8), 1953-1956 (2000-08-06)
Dehydroepiandrosterone (DHEA) and DHEA sulfate (DHEAS) may function as neurotrophic or neuroprotective factors to protect central nervous system (CNS) neurons against a variety of insults, including excitotoxicity. The present study evaluated the pharmacological effects of DHEAS in a reversible spinal
M Park-Chung et al.
Brain research, 830(1), 72-87 (1999-06-03)
Sulfated and unsulfated neurosteroids such as pregnenolone sulfate, dehydroepiandrosterone sulfate (DHEAS), pregnanolone, and allopregnanolone, modulate ionotropic amino acid neurotransmitter receptors, and may function as endogenous neuromodulators. The gamma-aminobutyric acid type A (GABAA) receptor exhibits both negative and positive modulation by

Articles

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

We offer many products related to GABAA receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service