Skip to Content
MilliporeSigma

Skip To

D5038

2′-Deoxyguanosine 5′-triphosphate trisodium salt solution

100 mM (pH 7)

Synonym(s):

dGTP-Na3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5Na3O13P3
CAS Number:
Molecular Weight:
573.13
NACRES:
NA.52
PubChem Substance ID:
UNSPSC Code:
41106305
EC Number:
300-026-5
MDL number:
Assay:
≥99%
Form:
liquid
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99%

Quality Segment

form

liquid

concentration

100 mM (pH 7)

color

colorless

foreign activity

DNase, RNase, none detected

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C(=O)N1

InChI

1S/C10H16N5O13P3.3Na/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(26-6)2-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h3-6,16H,1-2H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,11,13,14,17);;;/q;3*+1/p-3/t4-,5+,6+;;;/m0.../s1

InChI key

IWGGLKOTEOCWQP-BIHLCPNHSA-K

General description

2′-Deoxyguanosine 5′-triphosphate (dGTP) is made of a guanine nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar linked to a chain of three phosphate residues. It is used by the cells for the synthesis of DNA with the help of DNA polymerase.

Application

2′-Deoxyguanosine 5′-triphosphate trisodium salt is utilized for methods such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.
2′-Deoxyguanosine 5′-triphosphate trisodium salt solution has been used:

  • in a reverse transcription-polymerase chain reaction (RT-PCR) for DNA amplification.
  • as a component of the PCR mix for in situ reverse transcription-polymerase chain reaction (RT-PCR) amplification of mRNA from prokaryotic and eukaryotic ribosomes[1]
  • to study the mutations in the HBB gene via amplification refractory mutation system (ARMS)-PCR

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DGTP-ROD717011051466001
Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

-

form

liquid

form

solution

form

liquid

form

solution

assay

≥99%

assay

99% (dNTP)

assay

≥99%

assay

98%

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

concentration

100 mM (pH 7)

concentration

<0.9% (dNDP), 100 mM

concentration

10 mM in H2O

concentration

100 mM

color

colorless

color

-

color

colorless

color

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters