Skip to Content
MilliporeSigma

Skip To

DGTP-RO

Roche

dGTP

PCR Grade, sodium salt

Synonym(s):

2′-Deoxyguanosine 5′-triphosphate trisodium salt solution, dGTP-Na3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5Na3O13P3
CAS Number:
Molecular Weight:
573.13
UNSPSC Code:
12352302
PubChem Substance ID:
MDL number:
NACRES:
NA.54
Assay:
99% (dNTP)
Form:
solution
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99% (dNTP)

form

solution

usage

sufficient for 125,000 standard reactions (03732703001), sufficient for 31,250 standard reactions (11969030001), sufficient for 6,250 standard reactions (11934538001)

packaging

pkg of 1,250 μL (11969030001 [100 mM]), pkg of 4 × 1,240 μL (03732703001 [4x100 mM]), pkg of 250 μL (11934538001 [100 mM])

manufacturer/tradename

Roche

concentration

<0.9% (dNDP), 100 mM

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C(=O)N1

InChI

1S/C10H16N5O13P3.3Na/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(26-6)2-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h3-6,16H,1-2H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,11,13,14,17);;;/q;3*+1/p-3/t4-,5+,6+;;;/m0.../s1

InChI key

IWGGLKOTEOCWQP-BIHLCPNHSA-K

General description

2′-Deoxyguanosine 5′-triphosphate (dGTP) is made of a guanine nucleobase attached to deoxyribose and a 5′-hydroxyl on the sugar linked to a chain of three phosphate residues. It is used by the cells for the synthesis of DNA with the help of DNA polymerase.
dGTP, PCR Grade, is a 100 mM clear, colorless solution of the sodium salt (pH 8.3). PCR Grade nucleotides from Roche are specially manufactured and purified to the highest possible chemical purity. PCR Grade nucleotides contain at least 99% of the relevant deoxynucleoside-triphosphate (dNTP) and less than 0.9% deoxynucleoside-diphosphate (dNDP).

Application

2′-Deoxyguanosine 5′-triphosphate trisodium salt is utilized for methods such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.

Physical form

solution of dGTP in water (pH 8.3).

Preparation Note

Working solution: We recommend to prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).

Analysis Note

Function tested in RT-PCR. Tested for absence of contaminating deoxyribonucleases, ribonucleases and nicking activities according to the current Quality Control procedures.

Other Notes

For general laboratory use.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DNTP-RO11051466001DNSTP-RO
manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

form

solution

form

liquid

form

solution

form

liquid

assay

99% (dNTP)

assay

99% (dNTP)

assay

98%

assay

≥98% (dntp)

concentration

<0.9% (dNDP), 100 mM

concentration

<0.9% (dNDP)

concentration

100 mM

concentration

-

packaging

pkg of 1,250 μL (11969030001 [100 mM]), pkg of 250 μL (11934538001 [100 mM]), pkg of 4 × 1,240 μL (03732703001 [4x100 mM])

packaging

pkg of 4 × 1,250 μL (03622614001 [4x100 mM]), pkg of 4 × 250 μL (11969064001 [4x100 mM])

packaging

pkg of 250 μL (25 μmol; 100 mM)

packaging

pkg of 4 × 100 μL (11277049001 [4x10 μmol]), pkg of 40 × 100 μL (11922505001 [40x100mM])

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under dGTP


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library