Skip to Content
MilliporeSigma
All Photos(3)

Documents

D2157

Sigma-Aldrich

1,3-Dipalmitoyl-2-oleoylglycerol

≥99%

Synonym(s):

1,3-Dihexadecanoyl-2-(cis-9-octadecenoyl)glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H100O6
CAS Number:
Molecular Weight:
833.36
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C53H100O6/c1-4-7-10-13-16-19-22-25-26-29-32-35-38-41-44-47-53(56)59-50(48-57-51(54)45-42-39-36-33-30-27-23-20-17-14-11-8-5-2)49-58-52(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3/h25-26,50H,4-24,27-49H2,1-3H3/b26-25-

InChI key

FDCOHGHEADZEGF-QPLCGJKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Oxidative lipidomics to elucidate the non-volatile derivatives of four typical triglycerides in vegetable oils under simulated frying conditions: This research underscores the pivotal role of 1,3-Dipalmitoyl-2-oleoylglycerol in understanding the oxidative stability and decomposition products of triglycerides under high-heat conditions, relevant in food science and lipidomics (Hu et al., 2023).

  • Stearic acids at sn-1, 3 positions of TAG are more efficient at limiting fat deposition than palmitic and oleic acids in C57BL/6 mice: This research examines the impact of specific triglyceride configurations, including 1,3-Dipalmitoyl-2-oleoylglycerol, on lipid metabolism and fat accumulation in experimental models, which is crucial for understanding lipid-related diseases and for developing lipid-based therapeutics (Gouk et al., 2014).

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service