Skip to Content
MilliporeSigma
1,3-Dipalmitoyl-2-oleoylglycerol ≥99%

1,3-Dipalmitoyl-2-oleoylglycerol

1,3-Dipalmitoyl-2-oleoylglycerol ≥99%
Synonym(s):
1,3-Dihexadecanoyl-2-(cis-9-octadecenoyl)glycerol
Empirical Formula (Hill Notation):
C53H100O6
CAS Number:
Molecular Weight:
833.36

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing