Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D0781

Sigma-Aldrich

2,4-Diamino-6,7-diisopropylpteridine phosphate salt

≥98%

Synonym(s):

2,4-Diamino-6,7-diisopropylpteridine phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N6 · H3PO4
CAS Number:
Molecular Weight:
344.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

mp

232-234 °C (lit.)

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.CC(C)c1nc2nc(N)nc(N)c2nc1C(C)C

InChI

1S/C12H18N6.H3O4P/c1-5(2)7-8(6(3)4)16-11-9(15-7)10(13)17-12(14)18-11;1-5(2,3)4/h5-6H,1-4H3,(H4,13,14,16,17,18);(H3,1,2,3,4)

InChI key

YNQBQYASRYRNRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Diamino-6,7-diisopropylpteridine phosphate salt, is a phosphate salt of 2,4-diamino-6,7-diisopropylpteridine, which is a pteridine derivative. It is used as a valuable reagent in organic synthesis due to its ability to act as a nucleophile as well as an intermediate in various chemical reactions to synthesize heterocyclic compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrio cholerae resistant to 2, 4-diamino-6, 7-diisopropylpteridine (O/129) isolated from patients with enteritis in Cear{\'a}, Brazil
Hofer et al.
Journal of Health, Population, and Nutrition, 39-42 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service