Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H23N5O6
CAS Number:
Molecular Weight:
405.41
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
protect from light
Product Name
Roseoflavin, ≥95% (HPLC)
Quality Segment
assay
≥95% (HPLC)
form
powder
storage condition
protect from light
color
faint red to very dark brown-red
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
−20°C
SMILES string
[n]1(c2n[c]([nH][c](c2nc3c1cc(c(c3)C)N(C)C)=O)=O)CC(O)C(O)C(O)CO
InChI
1S/C18H23N5O6/c1-8-4-9-11(5-10(8)22(2)3)23(6-12(25)15(27)13(26)7-24)16-14(19-9)17(28)21-18(29)20-16/h4-5,12-13,15,24-27H,6-7H2,1-3H3,(H,21,28,29)
InChI key
IGQLDUYTWDABFK-UHFFFAOYSA-N
Biochem/physiol Actions
Roseoflavin is an analog of flavin mononucleotide (FMN) and riboflavin with antimicrobial activity.
Roseoflavin is an analog of flavin mononucleotide (FMN) and riboflavin with antimicrobial activity. Roseoflavin is converted to the flavin mononucleotide (FMN) analog roseoflavin mononucleotide (RoFMN) by flavokinase and to the flavin adenine dinucleotide (FAD) analog roseoflavin adenine dinucleotide (RoFAD) by FAD synthetase, and acts as a riboflavin antagonist. Roseoflavin is an inhibitor of bacterial riboflavin riboswitches, cis regulatory elements present in non-coding RNA that specifically bind to natural ligands to regulate gene expression. Roseoflavin, converted to RoFMN, blocks the flavin mononucleotide riboswitch-mediated expression of the ribB gene, which is required for riboflavin biosynthesis.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.