Skip to Content
MilliporeSigma

CRISPR11

CRISPR-Lenti Human Positive Control DNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41106609
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

General description

This control is intended for use as a positive control for knockout of human gene targets when used with clones of the Sigma lenti CRISPRs. The DNA format of this control construct permits direct transfection in knockout experiments or use in the viral packaging process (for lentiviral-based knockdown experiments).

Validated CRISPR site, which serves as an experimental control for the Sigma lenti CRISPRs. A one-component positive control system consisting of a EF1-alpha-driven Cas9, a U6-driven guide RNA plasmid targeting the human EMX1 gene, with both Puromycin and GFP co-expressed with Cas9. The Sigma lenti CRISPR Human EMX1 Positive Control Vector is a lentiviral-based plasmid vector that contains a guide RNA sequence targeting the human EMX1 (empty spiracles homeobox 1) gene. EMX1 belongs to the EMX homeobox family. The vector backbone is the same as the Sigma lenti CRISPR products.

Application

Functional Genomics/Screening/Target Validation
  • Positive control for creating gene knockouts/genetic modifications in cell lines.

To see more application data, protocols, vector maps visit sigma.com/crisprs.

Features and Benefits

Serves as an experimental control for the CRISPR editing workflow using lenti CRISPRs. Allows for validation of your system with the lenti CRISPR/Cas9 system. A positive result in a mis-match detection assay will indicate validation of your system.

Physical form

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively. Also, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP001). The human EMX1 positive Control Vector is provided as 1 ug of plasmid DNA in Tris-EDTA (TE).


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library