Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CPS3100

Sigma-Aldrich

Chemiluminescent Peroxidase Substrate-3

sufficient for 100 mL substrate

Synonym(s):

CPS3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

form

solution

Quality Level

usage

sufficient for 100 mL substrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

CPS3 formulations are economically priced and outperform the most commonly used competitors′ products. This two component system provides low nanogram detection with superior signal-to-noise ratio. In addition, the components are stable after mixing, adding to the flexibility of usage.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service