Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

07-328

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys8) Antibody

serum, Upstate®

Synonym(s):

H4K8Ac, Histone H4 (acetyl K8)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, Saccharomyces cerevisiae

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys8)

Gene Information

human ... H4C1(8359)

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.



The N-terminal tail of histone H4 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

Peptide corresponding to amino acids 3-13 of yeast Histone H4 (RGKGGAcKGLGKG-C) in which lysine 8 is acetylated.

Application

Use Anti-acetyl-Histone H4 (Lys8) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, IP, WB, DB, ChIP-seq to detect acetyl-Histone H4 (Lys8) also known as H4K8Ac, Histone H4 (acetyl K8).

Quality

routinely evaluated by immunoblot on acid extracts from sodium butyrate treated HeLa cells

Target description

10 kDa

Analysis Note

Control
Sodium butyrate treated HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cc Chemokine Ligand 2 and Lif Cooperatively Promote Pluripotency in Mouse Induced Pluripotent Cells.
Hasegawa Y, Takahashi N, Forrest AR, Shin JW, Kinoshita Y, Suzuki H, Hayashizaki Y
Stem Cells null
Gain-of-Function Activity of Mutant p53 in Lung Cancer through Up-Regulation of Receptor Protein Tyrosine Kinase Axl.
Vaughan, CA; Singh, S; Windle, B; Yeudall, WA; Frum, R; Grossman, SR; Deb, SP; Deb, S
Genes & Cancer null
Shivakumar Subbanna et al.
The international journal of neuropsychopharmacology, 18(5) (2015-01-23)
Ethanol exposure to rodents during postnatal day 7 (P7), which is comparable to the third trimester of human pregnancy, induces long-term potentiation and memory deficits. However, the molecular mechanisms underlying these deficits are still poorly understood. In the present study
Liposomes loaded with histone deacetylase inhibitors for breast cancer therapy.
Urbinati G, Marsaud V, Plassat V, Fattal E, Lesieur S, Renoir JM
International Journal of Pharmaceutics null
Negar Bagheri-Sereshki et al.
Biology of reproduction, 94(4), 81-81 (2016-02-26)
The coadministration of bleomycin, etoposide, and cisplatin (BEP) has increased the survival rate of testicular cancer patients to over 90%. Previous studies have demonstrated that BEP induces germ cell damage during the final stages of spermatogenesis, when major chromatin remodeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service