Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS4565

Corning® 1536 well plates, cyclic olefin co-polymer

black, non-treated surface, non-sterile

Synonym(s):

1536 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

black
black bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 10 ea
case of 50 ea

manufacturer/tradename

Corning 4565

well volume

12.5 μL

wells

1536 wells

working volume

0-8 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

1536 well cyclic olefin copolymer microplates are perfectly suited for high-throughput screening (HTS). A variety of surface treatments enable both cell-based and biochemical assays. The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols, allowing for compound storage as well.

Key Features
  • Cyclic Olefin Copolymer (COC) material
  • Flat top for improved sealing
  • 1536 Well Hi Base solid color microplates (black or white)
  • Variety of treatment options
  • tissue culture treated
  • Corning CellBIND® Surface
  • Nonbinding Surface (NBS)
  • Not treated
  • Bar coded (one label on both long sides)
  • Custom bar codes available for compatibility with the Kalypsys System
Key Benefits
  • Low autofluorescence
  • Broad chemical resistance including DMSO and alcohol, which allows for assay step reduction
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service