Skip to Content
MilliporeSigma

CLS3583

Corning® HTS Transwell® 96 well permeable support system, accessory plates

96 well receiver plate, TC-treated, flat bottom, sterile, w/lid, black, 10/cs

Synonym(s):

corning transwell, transwell corning

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
black polystyrene plate, flat bottom wells black , polystyrene
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black polystyrene plate, flat bottom wells black , polystyrene

sterility

sterile

feature

lid, skirt

packaging

case of 10, pack of 10

manufacturer/tradename

Corning 3583

technique(s)

cell culture: suitable

size

96 wells

color

black

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

General description

  • Product is an accessory for the Corning® HTS Transwell®-96 Permeable Support system
  • Plate is treated for optimal cell attachment
  • For use in cell growth or assay steps of an experiment
  • Used for fluorescent evaluation
  • When used with HTS Transwell®-96 unit, the plate allows for convenient pipette access to both apical and basolateral sides of each well (manual or automated)
  • Corner notched and SBS standard footprint
  • Sterile lid included with each plate

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library