Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS432022

Corning® CoolBox, XT System

workstation, single capacity, green

Synonym(s):

Staining, Staining Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300
NACRES:
NB.12
Pricing and availability is not currently available.

feature

workstation, single capacity

packaging

pkg of 1

manufacturer/tradename

Corning 432022

size

14.0 cm × 9.4 cm × 9.9 cm, Internal Dimensions
20.0 cm × 16.0 cm × 15.0 cm, External Dimensions

color

green

Looking for similar products? Visit Product Comparison Guide

General description

Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack and CoolSink thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.

Features and Benefits

Cooling with Lid Closed: 16 Hours
Cooling with Lid Open: 10 Hours
Freezing with Lid Closed: 8 Hours
Freezing with Lid Open: 5 Hours

Legal Information

CoolBox is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service