Key Documents
CLS431644
Corning® roller bottles, not treated
bottle surface area 850 cm2, Easy grip cap, polystyrene, sterile
Synonym(s):
Corning roller bottles
About This Item
UNSPSC Code:
41122105
NACRES:
NB.43
material
clear bottle
cylindrical bottle
high-density polyethylene cap
polystyrene
wide-mouth bottle
sterility
sterile
feature
cap
packaging
pack of 1 ea
case of 40 ea
manufacturer/tradename
Corning 431644
bottle neck diam.
~45 mm
capacity
255 mL
diam. × H
117 mm × 271 mm
binding type
non-treated surface
Features and Benefits
  • Caps are made of high-density polyethylene, whereas bottles are made of virgin polystyrene
  • Flexible tubing with heat sealability and chemically resistant
  • Seamless structure with printed graduations and lot number to help in product traceability
  • Easy grip cap with wide knurls for easy handling
  • Gamma radiation sterilized, having sterility assurance level (SAL) of 10-6
  • Nonpyrogenic
  • Compatible with disposable tubing set
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models