Key Documents
CLS431301
Corning® square bioassay dishes
low profile, without handles, not TC treated, square
Synonym(s):
cell culture dishes, cell culture plates, culture dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
Pricing and availability is not currently available.
material
polystyrene
square
sterility
sterile; γ-irradiated
manufacturer/tradename
Corning 431301
H
12.5 mm
L × W
245 mm × 245 mm
surface area
500 cm^2
General description
  • Corning® square bioassay dishes are made from optically clear polystyrene.
  • Stacking bead design facilitate secure stacking and prevents slipping.
  • Compatible with automated colony picking instruments.
  • Certified nonpyrogenic dishes are supplied with lids.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)