Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS354480

Corning® BioCoat Matrigel® 24 well plates

Tissue Culture (TC)-treated surface clear PET membrane (8.0um Pore Polyester (PET) Membrane), pkg of 24 ea (2 packs of 24), Invasion Chamber

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

Product Name

Corning® BioCoat Matrigel® Invasion Chamber, clear PET membrane (8.0um Pore Polyester (PET) Membrane), well size 24 wells

material

clear PET membrane (8.0um Pore Polyester (PET) Membrane)

feature

lid: no

packaging

pkg of 24 ea (2 packs of 24)

manufacturer/tradename

Corning 354480

well size

24 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service