Skip to Content
MilliporeSigma
All Photos(2)

Documents

CLS354265

Corning® Collagen I

from human(Natural), pack of 10 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.28

biological source

human (Natural)

packaging

pack of 10 mg

manufacturer/tradename

Corning 354265

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Corning Collagen I, human, 10mg, is effective as a gel or thin coating at a recommended concentration of 0.2-2μg/cm² of growth surface depending on cell type (guidelines for coating included with product).
Formulation
Frozen in 2mM HCl

Source
Human placenta. Source material tested for hepatitis B antigen and HIV-1 antibody.

Quality
Electrophoretically homogeneous (SDS-PAGE)
Shown to promote attachment and spreading of HT-1080 human fibrosarcoma cells
Found negative for bacteria, fungi, and mycoplasma

Preparation and Storage
Stable for at least three months at -20°C. Avoid multiple freeze-thaws.

Handling
Effective as a gel or thin coating at a recommended concentration of 0.2-2 μg/cm² of growth surface depending on cell type

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service