Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS354249

Corning® Collagen I

from rat(tail, High Protein), pack of 100 mg

Synonym(s):

Collagen I, Corning Collagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.7

biological source

rat (tail, High Protein)

packaging

pack of 100 mg

manufacturer/tradename

Corning 354249

Looking for similar products? Visit Product Comparison Guide

General description

Corning Collagen I HC, rat tail, 100 mg, is supplied as a liquid in 0.02N Acetic acid with a concentration range of 8-11 mg/ml. The product has been tested for its ability to promote the attachment and spreading of HT 1080 Human Fibrosarcoma cells. 1/pack, formerly Cat. #40040. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
Formulation
Collagen I HC, rat tail - 0.02N in Acetic Acid
Concentration range 8-11 mg/ml

Source
Rat tail tendon

Quality
Purity > 90% by SDS-PAGE
Tested for its ability to promote the attachment and spreading of HT 1080 Human Fibrosarcoma cells.
Found negative for the presence of bacteria, fungi and mycoplasma.
Forms a firm gel up to a concentration of 0.3 mg/ml. Further dilution may decrease the rigidity of the gel as will the time from manufacture.

Preparation and Storage
Stable for a minimum of 3 months from day of shipment when stored at 2° to 8°C. Do not freeze.

Handling
Rat Tail Collagen HC is used as a three dimensional gel and will form a firm gel up to a concentration of 0.3 mg/ml. Gelling procedures included with product.

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service