Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS354235

Corning® Dispase

pack of 100 mL

Synonym(s):

Dispase Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.7

packaging

pack of 100 mL

manufacturer/tradename

Corning 354235

General description

Corning Dispase, 100 mL, is a bacillus-derived neutral metalloprotease recommended for recovering cells cultured on Corning Matrigel Basement Membrane Matrix.
Molecular Weight
36kD

Source
Bacillus polymyxa-derived metallo protease

Quality
Tested for ability to solubilize a 1mm thick layer of gelled Corning Matrigel Matrix after two hours at 37°C
Tested and found negative for bacteria, fungi, and mycoplasma

Preparation and Storage
Stable for at least three months at -20°C. Avoid multiple freeze-thaws.

Handling
Recommended concentration - 10U/cm² of Corning Matrigel Matrix (e.g., 100U per 35mm dish).

Formulation
Frozen in HBSS, pH 7.4


This product is non-returnable. If you have any questions, please contact us immediately.

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service