Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS354086

Corning® BioCoat Poly-D-Lysine #1 German Glass Coverslip

size 12 mm, pkg of 80 ea (1 pack,Bulk Packed)

Synonym(s):

slides, coverslip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

glass

packaging

pkg of 80 ea (1 pack,Bulk Packed)

manufacturer/tradename

Corning 354086

size

12 mm

binding type

Tissue Culture (TC)-treated surface

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service