Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3392

Corning® HTS Transwell® 96 well permeable supports

HTS Transwell-96 Permeable Support with 1.0 μm pore polyester membrane, TC-treated, sterile, 5/cs

Synonym(s):

corning transwell, transwell corning

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.75
material:
flat bottom wells clear
polyethylene membrane
polystyrene
round wells
size:
96 wells

material

flat bottom wells clear
polyethylene membrane
polystyrene
round wells

sterility

sterile

feature

lid
skirt
plate format: 96 well

packaging

case of 5
pack of 5

manufacturer/tradename

Corning 3392

insert working volume

0.08 mL

membrane diam.

4.26 mm

membrane thickness

10.0 μm

size

96 wells

surface area

0.14 cm2 , cell growth area

well working volume

0.235 mL

color

clear plate

pore size

1.0 μm pore size

suitability

suitable for (cell culture applications; optimal for cell attachment)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Product includes Transwell® unit (1 integral tray with 96 wells), reservoir plate (for cell growth steps), receiver plate (96 wells for growth or assay steps) and 2 sterile lids; 5 sets per case
  • 1.0 μm pore size PET (polyester) membrane
  • Membrane is treated for optimal cell attachment
  • Cell growth area is 0.143 cm² which is 20-50% greater than other brands
  • Large apical and basolateral access ports
  • Automation optimized design with multi-channel feeder ports, improved gripping surface and bar coding

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service