Skip to Content
MilliporeSigma

CLS3385

Corning® HTS Transwell® 96 well permeable supports

HTS Transwell-96 Permeable Support with 3.0 μm pore polycarbonate membrane, TC-treated, sterile, 2/cs

Synonym(s):

corning transwell, transwell corning

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
flat bottom wells clear , polycarbonate membrane, polystyrene , round wells
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom wells clear , polycarbonate membrane, polystyrene , round wells

sterility

sterile

feature

lid, skirt, plate format: 96 well

packaging

pack of 1, case of 2

manufacturer/tradename

Corning 3385

technique(s)

cell culture: suitable

insert size

0.08 mL

membrane diam.

4.26 mm

membrane thickness

10.0 μm

size

96 wells

surface area

0.14 cm2 , cell growth area

well working volume

0.235 mL

color

clear plate

pore size

3.0 μm

suitability

suitable for (cell culture applications; optimal for cell attachment)

binding type

Tissue Culture (TC)-treated surface

General description

  • HTS Transwell®-96 permeable supports have an array of 96 wells with permeable inserts connected by a rigid robotics-friendly tray
  • Product includes HTS Transwell®-96 permeable support insert, receiver plate (96 wells for growth or assay steps) and 1 sterile lid; 2 sets per case
  • 3.0 μm pores size PC (polycarbonate) membrane
  • Membrane area for cell growth or cell migration is 0.143 cm² which is 20-50% greater than other brands
  • Large apical and basolateral access ports
  • Automation optimized design with multi-channel feeder ports, improved gripping surface and bar coding
  • All membranes are treated for cell attachment
  • Receiver plates are untreated
  • Lid – minimizes evaporation and protects against contamination

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library