Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3291

Corning® CellBIND® Surface cell culture flasks

CellBIND 150cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 50/cs

Synonym(s):

cell culture flasks, tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
eCl@ss:
32040501
NACRES:
NB.13

material

canted neck
cap (vented)
high-density polyethylene cap
rectangular bottom flask
rectangular flask
rectangular polystyrene
vent cap

sterility

sterile; γ-irradiated

feature

cap
graduations

packaging

pack of 5
case of 50

manufacturer/tradename

Corning 3291

maximum volume

210 mL

surface area

150 cm2

working volume

30-45 mL

suitability

suitable for (cell culture applications; optimal for adherent cell culturing)

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.

CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.

Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.

  • Manufactured from optically clear virgin polystyrene.
  • Treated CellBIND surface treatment for enhanced cell attachment
  • 100% integrity tested.
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service