Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human umbilical cord vein
Morphology:
(endothelial)
Pricing and availability is not currently available.
biological source
human umbilical cord vein
packaging
pkg of 1,000,000 cells
morphology
(endothelial)
application(s)
cell analysis (DNA, RNA, and protein)
shipped in
dry ice
storage temp.
−20°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.
Application
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord. They are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis. HUVEC are isolated from single donors in standard Endothelial Cell Growth Medium. (Human Umbilical Artery Endothelial Cells (HUAEC) from the same donor are available on request.)
Biochem/physiol Actions
Vein
Physical form
Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.
Preparation Note
Click here for more information.
Analysis Note
Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).
Other Notes
The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.
Legal Information
RNAlater is a registered trademark of Thermo Fisher Scientific, Inc.
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Still not finding the right product?
Explore all of our products under Human Umbilical Vein Endothelial Cells (HUVEC)
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.