Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS1395750

Pyrex® round media storage bottles and reusable screw caps

capacity 750 mL, case of 10 ea

Synonym(s):

glass bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
Pricing and availability is not currently available.

material

borosilicate glass bottle
cylindrical bottle
orange polypropylene cap (plug seal)

sterility

non-sterile

feature

cap
graduations
reusable

packaging

case of 10 ea

manufacturer/tradename

Corning 1395-750

availability

not available in EU

bottle neck diam.

30 mm

capacity

750 mL

fitting

GL 45

Looking for similar products? Visit Product Comparison Guide

General description

Pyrex heavy-duty round media storage bottles can be used for storage as well as for mixing and sampling. These media storage bottles Include linerless, one-piece, autoclavable, orange PP plug-seal caps with drip-free pouring rings. Optional red PBTP caps and red ETFE pouring rings are available separately, for dry heat sterilization (180°C).

Pyrex round media storage bottles are made of borosilicate glass with permanent, white enamel graduations and marking spots.

Caution: Bottles larger than 2 L should NOT be used with bottle-top filter units or in other applications involving vacuum/pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

A wide range of optional caps are available for these bottles:
  • Vented membrane cap with 0.22 μm PTFE hydrophobic membrane
  • Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C)
  • PBT caps with silicone septa for syringe sampling or introduction of reagents

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service